1,2,3,4-Tetrachlor-5,5-dimethoxycyclopentadien zur Synthese

Lieferant: Merck

Synonyme: 5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene, 1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene

8.14227.0005
8.14227.0005EA 0 EUR
8.14227.0005
1,2,3,4-Tetrachlor-5,5-dimethoxycyclopentadien zur Synthese
1,2,3,4-Tetrachlor-5,5-dimethoxycyclopentadien
Formel: C₇H₆Cl₄O₂
Molecular Weight: 263,93 g/mol
MDL: MFCD00001353
CAS-Nummer: 2207-27-4
EINECS: 218-623-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR