2-Methyl-2-thiazolin

Lieferant: Merck

Synonyme: 2-Methyl-4,5-dihydrothiazole

8.14232.0010
8.14232.0010EA 0 EUR
8.14232.0010
2-Methyl-2-thiazolin
2-Methyl-2-thiazolin
Formel: C₄H₇NS
Molecular Weight: 101.17 g/mol
MDL: MFCD00005314
CAS-Nummer: 2346-00-1
EINECS: 219-071-6

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point -101 °C
Molar Mass 101.17 g/mol
Boiling Point 144 - 146 °C (1013 hPa)
Flash Point 37 °C
Refractive Index 1.5211 (20 °C, 589 nm)
Density 1.08 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR