Methylphenylglyoxylat zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl benzoylformate, Methyl 2-oxo-2-phenylacetate

M-Clarity™ Qualitätsstufe = MQ200

8.14269.0025
8.14269.0025EA 97.5 EUR
8.14269.0025
Methylphenylglyoxylat zur Synthese, Sigma-Aldrich®
Methylphenylglyoxylat
Formel: C₉H₈O₃
Molecular Weight: 164,1600 g/mol
MDL: MFCD00008443
CAS-Nummer: 15206-55-0
EINECS: 239-263-3

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Melting Point 16 °C
Molar Mass 164.16 g/mol
Boiling Point 137 °C (19 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 77 °C
Refractive Index 1.585 (20 °C, 589 nm)
Density 1.16 g/cm3 (20 °C)
Ignition Point 500 °C
Synonyms Phenylglyoxylic acid methyl ester, Benzoyl formic acid methyl ester
Hill Formula C9H8O3
Chemical Formula C6H5COCOOCH3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR