tert-Butyl(2-nitroethyl)ether zur Synthese, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

8.14278.0001
8.14278.0001EA 108 EUR
8.14278.0001
tert-Butyl(2-nitroethyl)ether zur Synthese, Sigma-Aldrich®
tert-Butyl(2-nitroethyl)ether
Formel: C₆H₁₃NO₃
CAS-Nummer: 77791-00-5

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Molar Mass 147.17 g/mol
Boiling Point 30 - 35 °C (1 hPa)
Flash Point 75 °C
Density 1.00 g/cm3 (20 °C)
Synonyms 2-Nitroethanol tert-butyl ether
Hill Formula C6H13NO3
Chemical Formula O2NCH2CH2OC(CH3)3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR