(2-Chlorethoxy)benzol

Lieferant: Merck

8.14338.0100
8.14338.0100EA 0 EUR
8.14338.0100
(2-Chlorethoxy)benzol
(2-Chlorethoxy)benzol
Formel: C₆H₅OCH₂CH₂Cl
MDL: MFCD00018946
CAS-Nummer: 622-86-6
EINECS: 210-757-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR