(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamin zur Synthese

Lieferant: Merck

8.14399.0001
8.14399.0001EA 0 EUR
8.14399.0001
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamin zur Synthese
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamin
Formel: C₁₄H₁₉FeN
MDL: MFCD00066273
CAS-Nummer: 31886-58-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR