2-(Trimethylsilyl)thiazol

Lieferant: Merck

8.14407.0001
8.14407.0001EA 0 EUR
8.14407.0001
2-(Trimethylsilyl)thiazol
2-(Trimethylsilyl)thiazole
Formel: C₆H₁₁NSSi
MDL: MFCD00066274
CAS-Nummer: 79265-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR