α,α-Dichlorpropiophenon

Lieferant: Merck

8.14489.0005
8.14489.0005EA 0 EUR
8.14489.0005
α,α-Dichlorpropiophenon
α,α-Dichlorpropiophenon
Formel: C₆H₅C(O)CCl₂CH₃
Molecular Weight: 203,07 g/mol
Siedepunkt: 110…114 °C (19 hPa)
Dichte: 1,26 g/cm³ (20 °C)
CAS-Nummer: 57169-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR