2,2-Dichlorbutyrophenon zur Synthese
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.14490.0005
8.14490.0005EA
0
EUR
8.14490.0005
2,2-Dichlorbutyrophenon zur Synthese
2,2-Dichlorbutyrophenon
Formel:
C₁₀H₁₀Cl₂O |
CAS-Nummer:
66255-85-4 |
Spezifikation Testergebnisse
Molar Mass | 217.09 g/mol |
Boiling Point | 132 - 136 °C |
Density | 1.23 g/cm3 (20 °C) |
Synonyms | 2,2-Dichloropropyl phenyl ketone |
Hill Formula | C10H10Cl2O |
Chemical Formula | C6H5COC(Cl2)CH2CH3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...