Dimethyl adipate : dimethyl glutarate : dimethyl succinate (16:63:21) (Gemisch aus den Estern) zur Synthese, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Imsol
M-Clarity™ Qualitätsstufe = MQ200
8.14499.1000
8.14499.1000EA
47.7
EUR
8.14499.1000
Dimethyl adipate : dimethyl glutarate : dimethyl succinate (16:63:21) (Gemisch aus den Estern) zur Synthese, Sigma-Aldrich®
Dimethyl adipate : dimethyl glutarate : dimethyl succinate
Spezifikation Testergebnisse
Assay complete test mixture (GC, area%) | ≥ 98.0 % (a/a) |
Density (d 20 °C/ 4 °C) | 1.086 - 1.090 |
Identity (IR) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...