(±)-Campherchinon zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone
M-Clarity™ Qualitätsstufe = MQ200
8.14531.0010
8.14531.0010EA
158
EUR
8.14531.0010
(±)-Campherchinon zur Synthese, Sigma-Aldrich®
(±)-Campherchinon
Formel:
C₁₀H₁₄O₂ Molecular Weight: 166.22 g/mol |
MDL:
MFCD00064160 CAS-Nummer: 10373-78-1 EINECS: 233-814-1 |
Spezifikation Testergebnisse
Solubility | (20 °C) slightly soluble |
Melting Point | 198 - 201 °C |
Molar Mass | 166.22 g/mol |
Synonyms | 2,3-Bornanedione |
Hill Formula | C10H14O2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...