3-Methoxy-2-cyclohexen-1-on zur Synthese
Lieferant: Merck
Synonyme:
3-Methoxycyclohex-2-enone
M-Clarity™ Qualitätsstufe = MQ200
8.14561.0010
8.14561.0010EA
111
EUR
8.14561.0010
3-Methoxy-2-cyclohexen-1-on zur Synthese
3-Methoxy-2-cyclohexen-1-on
Formel:
C₇H₁₀O₂ Molecular Weight: 126,16 g/mol |
MDL:
MFCD00236366 CAS-Nummer: 16807-60-6 EINECS: 240-837-0 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 32 - 35 °C |
Molar Mass | 126.16 g/mol |
Boiling Point | 114 °C (19 hPa) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...