N-Acetyl-DL-Valin zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Acetamido-3-methylbutanoic acid, Ac-DL-Val-OH
M-Clarity™ Qualitätsstufe = MQ200
8.14599.0050
8.14599.0050EA
84.3
EUR
8.14599.0050
N-Acetyl-DL-Valin zur Synthese, Sigma-Aldrich®
N-Acetyl-DL-Valin
Formel:
C₇H₁₃NO₃ Molecular Weight: 159.2 g/mol |
MDL:
MFCD00066065 CAS-Nummer: 3067-19-4 EINECS: 221-321-4 |
Spezifikation Testergebnisse
Solubility | 37 g/l (25 °C) |
Melting Point | 146 - 149 °C |
Molar Mass | 159.18 g/mol |
Hill Formula | C7H13NO3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...