Formamidindisulfiddihydrochlorid zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dithiourea Dihydrochloride, Dithiodiformamide Dihydrochloride

M-Clarity™ Qualitätsstufe = MQ200

8.14609.0010
8.14609.0010EA 96.8 EUR
8.14609.0010
Formamidindisulfiddihydrochlorid zur Synthese, Sigma-Aldrich®
Formamidine disulphide dihydrochloride
Formel: C₂H₆N₄S₂
Molecular Weight: 223,13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00012586
CAS-Nummer: 14807-75-1
EINECS: 238-876-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a806fc5b7d63893d1e4d70ee4d7a837bd04a0e5f&vid=a806fc5b7d63893d1e4d70ee4d7a837bd04a0e5f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 175 - 179 °C
Molar Mass 223.15 g/mol
Storage +2...+8 °C
Assay (argentometric) ≥97,0% (m)
Identity Conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR