Ethylendiammoniumdiiodid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.14619.0025
8.14619.0025EA
59.9
EUR
8.14619.0025
Ethylendiammoniumdiiodid, Sigma-Aldrich®
Ethylendiammoniumdiiodid
Formel:
C₂H₁₀I₂N₂ |
MDL:
MFCD00035535 CAS-Nummer: 5700-49-2 EINECS: 227-186-8 |
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Molar Mass | 315.92 g/mol |
pH | 4.0 - 6.0 (100 g/l, H2O, 20 °C) |
Hill Formula | C2H10I2N2 |
Chemical Formula | C2H8N2*2HI |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...