4,4'-Methylenbis(3-chlor-2,6-diethylanilin) zur Synthese, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

8.14676.0025
8.14676.0025EA 41.8 EUR
8.14676.0025
4,4'-Methylenbis(3-chlor-2,6-diethylanilin) zur Synthese, Sigma-Aldrich®
4,4'-Methylenbis(3-chlor-2,6-diethylanilin)
Formel: C₂₁H₂₈Cl₂N₂
Molecular Weight: 379,3664 g/mol
MDL: MFCD00071551
CAS-Nummer: 106246-33-7
EINECS: 402-130-7
REACH: 01-0000015192-78

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) slightly soluble
Melting Point 88 - 90 °C
Molar Mass 379.37 g/mol
Bulk Density 610 - 650 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR