3,3'-Diaminodiphenylsulfon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,3'-Sulfonyldianilin, DADPS, 3-Aminophenylsulfon, Bis(3-aminophenyl)sulfon

M-Clarity™ Qualitätsstufe = MQ200

8.14680.0025
8.14680.0025EA 47.6 EUR
8.14680.0025
3,3'-Diaminodiphenylsulfon, Sigma-Aldrich®
3,3'-Diaminodiphenylsulfon
Formel: C₁₂H₁₂N₂O₂S
MDL: MFCD00007792
CAS-Nummer: 599-61-1
EINECS: 209-967-5

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) slightly soluble
Melting Point 167 - 170 °C
Molar Mass 248.29 g/mol
Bulk Density 400 kg/m3
Synonyms 3,3'-Sulfonyldianiline
Hill Formula C12H12N2O2S


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR