Hexadecyltrimethylammonium-p-toluenesulphonate zur Synthese, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
N,N,N-Trimethylhexadecan-1-aminium 4-methylbenzenesulphonate
M-Clarity™ Qualitätsstufe = MQ200
8.14692.0100
8.14692.0100EA
95.2
EUR
8.14692.0100
Hexadecyltrimethylammonium-p-toluenesulphonate zur Synthese, Sigma-Aldrich®
Hexadecyltrimethylammonium-p-toluenesulphonate
Formel:
C₂₆H₄₉NO₃S Molecular Weight: 455,742 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061383 CAS-Nummer: 138-32-9 EINECS: 205-324-8 UN: 2811 ADR: 6.1,II |
Spezifikation Testergebnisse
Solubility | 10 g/l (25 °C) |
Melting Point | 245 - 250 °C |
Molar Mass | 455.74 g/mol |
pH | 6 - 8 (H2O, 20 °C) (saturated aqueous solution) |
Synonyms | Cetyltrimethylammonium tosylate, Cetats, Tricetol |
Hill Formula | C26H49NO3S |
Identity | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...