2-tert-Butyl-4-methylcyclohexanol zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Rootanol, 4-methyl-2-t-butylcyclohexanol, 2-(1,1-dimethylethyl)-4-methylcyclohexan-1-ol

M-Clarity™ Qualitätsstufe = MQ200

8.14694.0025
8.14694.0025EA 54 EUR
8.14694.0025
2-tert-Butyl-4-methylcyclohexanol zur Synthese, Sigma-Aldrich®
2-tert-Butyl-4-methylcyclohexanol
Formel: C₁₁H₂₂O
Molecular Weight: 170,3 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 67634-11-1
EINECS: 266-815-0

Bestellen

Spezifikation Testergebnisse

Solubility Slightly soluble (20 °C)
Molar Mass 170.3 g/mol
Boiling Point 215 - 216 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 104 °C
Explosion Limit 1.4 - 6.6 %(V)
Density 0.9 g/cm3 (20 °C)
Ignition Point 275 °C
Melting point +112...+113 °C
Vapour pressure -1 hPa (20 °C)
Assay isomers (GC, area%) ≥97% (a/a)
Identity (IR) Passes test
Storage temperature +15...+25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR