4,5-Diamino-6-hydroxy-2-pyrimidinethiol hemisulphate-1,5-hydrate zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Bis(6-amino-4-oxo-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinaminium) sulphate, 4(1H)-Pyrimidinone, 5,6-diamino-2,3-dihydro-2-thioxo-, sulphate (2:1)
M-Clarity™ Qualitätsstufe = MQ200
8.14702.0005
8.14702.0005EA
77.8
EUR
8.14702.0005
4,5-Diamino-6-hydroxy-2-pyrimidinethiol hemisulphate-1,5-hydrate zur Synthese, Sigma-Aldrich®
4,5-Diamino-6-hydroxy-2-pyrimidinethiol hemisulphate-1,5-hydrate
Formel:
C₈H₁₄N₈O₆S₃ Molecular Weight: 414,442 g/mol |
CAS-Nummer:
207226-25-3 |
Spezifikation Testergebnisse
Melting Point | 235 °C |
Hill Formula | C8H14N8O6S3*(H2O)n |
Assay | ≥97,0% (a/a) |
Identity | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...