4-tert-Butylcalix[4]aren zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
p-tert-Butylcalix[4]arene
M-Clarity™ Qualitätsstufe = MQ200
8.14707.0001
8.14707.0001EA
118
EUR
8.14707.0001
4-tert-Butylcalix[4]aren zur Synthese, Sigma-Aldrich®
4-tert-Butylcalix[4]aren
Formel:
C₄₄H₅₆O₄ Molecular Weight: 648.93 g/mol |
MDL:
MFCD00066280 CAS-Nummer: 60705-62-6 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 344 - 347 °C |
Molar Mass | 648.92 g/mol |
Hill Formula | C44H56O4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...