Dimethoxy(diisobutyl)silan zS, Sigma-Aldrich®
Lieferant: Merck
|
|
Synonyme:
Diisobutyldimethoxysilane
MQ200
8.14720.0025
8.14720.0100
8.14720.0025EA
65.3
EUR
8.14720.0025
8.14720.0100
Dimethoxy(diisobutyl)silan zS, Sigma-Aldrich®
Dimethoxy(diisobutyl)silan
Formel:
C₁₀H₂₄O₂Si Molecular Weight: 204.38 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236510 CAS-Nummer: 17980-32-4 UN: 3082 ADR: 9 III |
Spezifikation Testergebnisse
Solubility | 13.4 mg/l (25 °C) Hydrolysis |
Molar Mass | 204.38 g/mol |
Boiling Point | 188 °C (1025 hPa) |
Vapor Pressure | 0.75 hPa (25 °C) |
Flash Point | 62 °C |
Density | 0.863 g/cm3 (20 °C) |
Ignition Point | 255 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...