Natrium-D(+)-glucuronat Monohydrat zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: D-Glucuronic acid sodium salt hydrate, D-Glucuronic acid sodium salt monohydrate, D(+)-Glucuronic acid, sodium salt, monohydrate, Sodium (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate

M-Clarity™ Qualitätsstufe = MQ200

8.14723.0010
8.14723.0010EA 58.5 EUR
8.14723.0010
Natrium-D(+)-glucuronat Monohydrat zur Synthese, Sigma-Aldrich®
Natrium-D(+)-glucuronat Monohydrat
Formel: C₆H₁₀O₇·1H₂O
Molecular Weight: 234,14 g/mol (216,14 anhy)
MDL: MFCD00151051
CAS-Nummer: 207300-70-7
EINECS: 239-065-7

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 137 - 141 °C
Molar Mass 234.14 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR