Diethylammoniumbromid zur Synthese, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
8.14863.0250
8.14863.0050
8.14863.0250EA
121
EUR
8.14863.0250
8.14863.0050
Diethylammoniumbromid zur Synthese, Sigma-Aldrich®
Diethylammoniumbromid
Formel:
C₄H₁₂BrN Molecular Weight: 154.05 g/mol |
MDL:
MFCD00012498 CAS-Nummer: 6274-12-0 EINECS: 228-466-2 |
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Melting Point | 215 - 218 °C |
Molar Mass | 154.05 g/mol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...