3-Methylbutylisovalerat zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Isovaleric acid Isopentyl ester, Isovaleric acid Isoamyl ester, Isoamyl Isovalerate, Isopentyl 3-methylbutanoate, Isopentyl Isovalerate

M-Clarity™ Qualitätsstufe = MQ200

8.14993.0250
8.14993.0250EA 69.9 EUR
8.14993.0250
3-Methylbutylisovalerat zur Synthese, Sigma-Aldrich®
3-Methylbutylisovalerat
Formel: C₁₀H₂₀O₂
Molecular Weight: 172,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00048369
CAS-Nummer: 659-70-1
EINECS: 211-536-1
UN: 3082
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Solubility 0.016 g/l (20 °C)
Molar Mass 172.27 g/mol
Boiling Point 195 °C (1013 hPa)
Vapor Pressure 0.8 hPa (20 °C)
Flash Point 80 °C
Density 0.855 g/cm3 (25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR