Ammoniumacetat-[D7]

Lieferant: Merck

Synonyme: Essigsäure Ammoniumsalz-D7, (2H3)Essigsäure (2H4)Ammoniumsalz, (2H7)Essigsäure Ammoniumsalz, (2H7)Ammoniumacetat, (2H4)Ammonium(2H3)acetat

8.15010.0005
8.15010.0005EA 0 EUR
8.15010.0005
Ammoniumacetat-[D7]
Ammoniumacetat-[D7]
Formel: D₃CCOOND₄
Molecular Weight: 84,03 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00190378
CAS-Nummer: 194787-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR