Deuteriumbromid 48% in D₂O (99% D) (Isotop)

Lieferant: Merck

Synonyme: Bromwasserstoff-D1, Hydrogenbromid-D1

8.15015.0010
8.15015.0010EA 0 EUR
8.15015.0010
Deuteriumbromid 48% in D₂O (99% D) (Isotop)
Deuteriumbromid
Formel: DBr
Molecular Weight: 81,9 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044232
CAS-Nummer: 13536-59-9
EINECS: 236-894-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR