Hexamethylphosphoramid-[D18] Isotope_(99.5% D) NMR

Lieferant: Merck

Synonyme: Hexamethylphosphoramide-[²H₁₈]

8.15042.0005
8.15042.0005EA 0 EUR
8.15042.0005
Hexamethylphosphoramid-[D18] Isotope_(99.5% D) NMR
Hexamethylphosphoramid-[D18]
Formel: C₆D₁₈N₃OP
CAS-Nummer: 51219-90-0
EINECS: 257-063-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR