2-Propanol-[D7]

Lieferant: Merck
Gefahr

8.15046.0001
8.15046.0001EA 0 EUR
8.15046.0001
2-Propanol-[D7]
2-Propanol-[D7]
Formel: D₃CCD(OD)CD₃
Molecular Weight: 67.04 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 19214-96-1
UN: 1219
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR