2,4-Dichinolindicarbonsäure zS, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Quinoline-2,4-dicarboxylic acid
MQ200
8.15103.0025
8.15103.0025EA
200
EUR
8.15103.0025
2,4-Dichinolindicarbonsäure zS, Sigma-Aldrich®
2,4-Diquinolinecarboxylic acid
Formel:
C₁₁H₇NO₄ |
CAS-Nummer:
5323-57-9 |
Spezifikation Testergebnisse
Melting Point | 229 °C |
Molar Mass | 217.18 g/mol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...