Polyethylene glycol 400, EMPROVE® ESSENTIAL JP, Ph. Eur. geeignet für die Verwendung als Excipient, SAFC®

Lieferant: Merck

Synonyme: PEG 400, Polyethylenoxid 400, Polyoxyethylen 400, Polyglycol 400

8.17003.5000 8.17003.9220 8.17003.9025
8.17003.5000EA 461 EUR
8.17003.5000 8.17003.9220 8.17003.9025
Polyethylene glycol 400, EMPROVE® ESSENTIAL JP, Ph. Eur. geeignet für die Verwendung als Excipient, SAFC®
Polyethylenglycol 400

Other residual solvents acc. to ICH-guideline are excluded by the manufacturing process.
Residues of metal catalysts or metal reagents acc. to EMEA/CHMP/SWP/4446/2000 are not likely to be present.
Corresponds to Ph Eur, JP

Formel: H(OCH₂CH₂)nOH
Molecular Weight: 400 g/mol
Siedepunkt: ∼250 °C (1013 hPa)
Schmelzpunkt: 4…8 °C
Dichte: 1.13 g/cm³ (20 °C)
Flammpunkt: 245
Lagertemperatur: Raumtemperatur
MDL: MFCD00081839
CAS-Nummer: 25322-68-3
EINECS: 500-038-2

Bestellen

Spezifikation Testergebnisse

Identity (according to JP) passes test
Identity (according to Ph. Eur.) passes test
Appearance clear, viscous, colourless hygroscopic liquid
Appearance of solution passes test
Acidity,Alkalinity passes test
Reducing substances passes test
pH-value (5 %; water, 25 °C) 4.0 - 7.0
Density (d 20/20) 1.110 - 1.140
Viscosity (20 °C) 100 - 111 mm²/s
Viskosity (20 °C) 112 - 124 mPa∙s
Heavy metals ≤ 20 ppm
Formaldehyde ≤ 30 ppm
Ethylene glycol and diethylene glycol ≤ 0.25 %
Ethylene glycol (acc. to ICH-guideline class 2) ≤ 620 ppm
Ethylene oxide (GC) ≤ 1 ppm
Dioxane (GC) ≤ 10 ppm
Hydroxyl value 264 - 300
Average molecular mass 380 - 420
Sulfated ash ≤ 0.1 %
Water (K. F.) ≤ 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR