Polyethylene glycol 4.000, Pulver, EMPROVE® ESSENTIAL Ph. Eur., SAFC®
Lieferant: Merck
8.17006.5000
8.17006.5000EA 465 EUR
8.17006.5000
Polyethylene glycol 4.000, Pulver, EMPROVE® ESSENTIAL Ph. Eur., SAFC®
Polyethylenglycol 4.000
Formel: H(OCH₂CH₂)nOH
Siedepunkt: ∼250 °C (1013 hPa)
Schmelzpunkt: 53…58 °C
Dichte: 1,13 g/cm³ (20 °C)
Flammpunkt: 250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01779612
CAS-Nummer: 25322-68-3
EINECS: 500-038-2
Bestellen
Spezifikation Testergebnisse
Identity passes test
Appearance white or almost white solid with a waxy or paraffin-like appearance
Appearance of solution passes test
Acidity,Alkalinity passes test
Reducing substances passes test
Density (d 20/20) about 1.080 (50% solution)
Solidification point 53 - 59 °C
Viscosity (20 °C) 106 - 131 mm²/s
Viskosity (20 °C) 114 - 142 mPa∙s
Formaldehyde ≤ 30 ppm
Ethylene oxide (GC) ≤ 1 ppm
Dioxane (GC) ≤ 10 ppm
Hydroxyl value 25 - 32
Average molecular mass 3700 - 4400
Sulfated ash ≤ 0.2 %
Water (K. F.) ≤ 1.0 %
Other residual solvents acc. to ICH-guideline are excluded by the manufacturing process.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not
likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur