1,3,5-Tribenzylhexahydro-1,3,5-triazin zur Synthese

Lieferant: Merck

8.18024.0025
8.18024.0025EA 0 EUR
8.18024.0025
1,3,5-Tribenzylhexahydro-1,3,5-triazin zur Synthese
1,3,5-Tribenzylhexahydro-1,3,5-triazin
Formel: C₂₄H₂₇N₃
MDL: MFCD00014599
CAS-Nummer: 2547-66-2
EINECS: 219-831-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR