1,3,5-Tributylhexahydro-1,3,5-triazin zur Synthese

Lieferant: Merck

8.18039.0025
8.18039.0025EA 0 EUR
8.18039.0025
1,3,5-Tributylhexahydro-1,3,5-triazin zur Synthese
1,3,5-Tributylhexahydro-1,3,5-triazin
Formel: C₁₅H₃₃N₃
CAS-Nummer: 13036-83-4
EINECS: 235-905-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR