1,3-Dimethyl-2-imidazolidinon ≥99,0% (durch GC) zur Synthese, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
1,3-Dimethylimidazolidin-2-on
M-Clarity™ Qualitätsstufe = MQ200
8.18212.6190
8.18212.6190EA
0
EUR
8.18212.6190
1,3-Dimethyl-2-imidazolidinon ≥99,0% (durch GC) zur Synthese, Sigma-Aldrich®
1,3-Dimethyl-2-imidazolidinon
Formel:
C₅H₁₀N₂O Molecular Weight: 114,15 g/mol |
MDL:
MFCD00003188 CAS-Nummer: 80-73-9 |
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Melting Point | 8.2 °C |
Molar Mass | 114.14 g/mol |
Boiling Point | 104 °C (20 hPa) |
Vapor Pressure | < - 1 hPa (20 °C) |
Flash Point | 114 °C |
Refractive Index | 1.472 (20 °C, 589 nm) |
Explosion Limit | 1.3 - 8.4 %(V) |
Density | 1.056 g/cm3 (20 °C) |
pH | (H2O) neutral |
Ignition Point | 305 °C |
Synonyms | Dimethylethylene urea, DMI, DMEU |
Hill Formula | C5H10N2O |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...