Glycin-tert-butylester (H-Gly-OtBu) zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-Butyl 2-aminoacetate, tert-Butyl Glycinate

M-Clarity™ Qualitätsstufe = MQ200

8.18213.0006
8.18213.0006EA 117 EUR
8.18213.0006
Glycin-tert-butylester (H-Gly-OtBu) zur Synthese, Sigma-Aldrich®
Glycin-tert-butylester (H-Gly-OtBu)
Formel: C₆H₁₃NO₂
Molecular Weight: 131,18 g/mol
MDL: MFCD00038194
CAS-Nummer: 6456-74-2

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Molar Mass 131.17 g/mol
Boiling Point 29 - 31 °C (3 hPa)
Refractive Index 1.4237 (20 °C, 589 nm)
Density 0.96 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR