(2R,6S)-(+)-4-Benzoyloxy-2-benzoyloxymethyl-6-methoxy-2H-pyran-3(6H)-one zS
Lieferant: Merck
MQ200
8.18233.0001
8.18233.0001EA
0
EUR
8.18233.0001
(2R,6S)-(+)-4-Benzoyloxy-2-benzoyloxymethyl-6-methoxy-2H-pyran-3(6H)-one zS
(2R,6S)-(+)-4-Benzoyloxy-2-benzoyloxymethyl-6-methoxy-2H-pyran-3(6H)-one
Formel:
C₂₁H₁₈O₇ Molecular Weight: 382.368 g/mol |
CAS-Nummer:
25552-06-1 |
|
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 124 - 127 °C |
Molar Mass | 382.37 g/mol |
Synonyms | Methyl-3,6-dibenzoyl-2-deoxy-α-D-glycero-hex-2-enopyranosid-4-ulose |
Hill Formula | C21H18O7 |
Storage | +15...+25 °C |
Assay (HPLC, area%) | ≥99,0% (a/a) |
Specific rotation | 70,0 - 71,0 ° |
Identity (IR) | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...