α-Brompropiophenon
Lieferant: Merck
Synonyme:
2-Brompropiophenon
8.18240.0025
8.18240.0025EA
0
EUR
8.18240.0025
α-Brompropiophenon
α-Brompropiophenon
Formel:
C₆H₅C(O)CHBrCH₃ Molecular Weight: 213,07 g/mol Siedepunkt: 245 °C (1013 hPa) Dichte: 1,439 g/cm³ (0 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000145 CAS-Nummer: 2114-00-3 EINECS: 218-307-5 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...