(+)-Ethyl-4,6-di-O-acetyl-2,3-dideoxy-α-D-erythro-hex-2-enopyranoside zur Synthese

Lieferant: Merck

Synonyme: 3beta-Acetoxy-2alpha-(acetoxymethyl)-6beta-ethoxy-3,6-dihydro-2H-pyran, (+)-Ethyl-4,6-di-O-acetyl-2,3-dideoxyalpha-D-erythro-hex-2-eno-pyranoside

M-Clarity™ Qualitätsstufe = MQ200

8.18289.0001
8.18289.0001EA 0 EUR
8.18289.0001
(+)-Ethyl-4,6-di-O-acetyl-2,3-dideoxy-α-D-erythro-hex-2-enopyranoside zur Synthese
(+)-Ethyl-4,6-di-O-acetyl-2,3-dideoxy-α-D-erythro-hex-2-enopyranoside
Formel: C₁₂H₁₈O₆
Molecular Weight: 258,27 g/mol
MDL: MFCD01103303
CAS-Nummer: 3323-72-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb239bd62a4e260803d26d608f21be424e680679&vid=bb239bd62a4e260803d26d608f21be424e680679&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Melting Point 76 - 78 °C
Molar Mass 258.27 g/mol
Bulk Density 200 kg/m3
Hill Formula C12H18O6
Storage +15...+25 °C
Assay (DSC) ≥99,0%
Specific rotation 105,0 - 107,0 °
Identity (IR) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR