Imidazol-4,5-dicarbonsäure zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1H-Imidazole-4,5-dicarboxylic acid, 4,5-Imidazoledicarboxylic acid
M-Clarity™ Qualitätsstufe = MQ200
8.18370.0010
8.18370.0010EA
95.2
EUR
8.18370.0010
Imidazol-4,5-dicarbonsäure zur Synthese, Sigma-Aldrich®
Imidazol-4,5-dicarbonsäure
Formel:
C₅H₄N₂O₄ Molecular Weight: 156,10 g/mol |
MDL:
MFCD00005200 CAS-Nummer: 570-22-9 EINECS: 209-327-5 |
Spezifikation Testergebnisse
Solubility | 0.5 g/l (20 °C) |
Melting Point | 270 - 275 °C |
Molar Mass | 156.1 g/mol |
Bulk Density | 650 - 660 kg/m3 |
pH | 3.7 (0.5 g/l, H2O, 20 °C) |
Ignition Point | 430 °C |
Hill Formula | C5H4N2O4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...