(S)-(+)-Carvon, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
D(+)-Carvon, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-on, D-p-Mentha-1(6),8-dien-2-on
M-Clarity™ Qualitätsstufe = MQ200
8.18410.0025
8.18410.0025EA
51.1
EUR
8.18410.0025
(S)-(+)-Carvon, Sigma-Aldrich®
(S)-(+)-Carvon
Formel:
C₁₀H₁₄O Molecular Weight: 150,22 g/mol Siedepunkt: 230 °C (1013 hPa) Schmelzpunkt: ? 15 °C Dichte: 0,9584 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00062997 CAS-Nummer: 2244-16-8 EINECS: 218-827-2 |
Spezifikation Testergebnisse
Hill Formula | C₁₀H₁₄O |
Molar Mass | 150.22 g/mol |
Boiling point | 228 - 230 °C (1013 hPa) |
Density | 0.96 g/cm³ (20 °C) |
Flash point | 88 °C DIN 51758 |
Refractive Index | 1.4988 (20 °C, 589 nm) |
Solubility | 1300 mg/l (18 °C) |
LD 50 oral | LD50 Rat 3562 mg/kg |
Storage class | 10 Combustible liquids not in Storage Class 3 |
WGK | WGK 1 slightly water endangering |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...