2,2'-Methylenebis(6-tert-butyl-p-cresol) zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2,2'-Methylenebis(4-methyl-6-tert-butylphenol), 2,2-Methylenebis(6-tert-butyl-p-cresol), Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, 2,2'-Methylenebis(6-tert-butyl-4-methylphenol), 3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5'-dimethyldiphenylmethane

M-Clarity™ Qualitätsstufe = MQ200

8.18447.9045
8.18447.9045EA 0 EUR
8.18447.9045
2,2'-Methylenebis(6-tert-butyl-p-cresol) zur Synthese, Sigma-Aldrich®
2,2'-Methylenebis(6-tert-butyl-p-cresol)
Formel: C₂₃H₃₂O₂
Molecular Weight: 340.51 g/mol
MDL: MFCD00043641
CAS-Nummer: 119-47-1

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) practically insoluble
Melting Point 125 - 128 °C
Molar Mass 340.5 g/mol
Bulk Density 300 kg/m3
Boiling Point 187 °C (0.7 hPa)
Flash Point 195 °C
Density 1.07 g/cm3 (20 °C)
pH 4 - 7 (H2O, 20 °C) (aqueous suspension)
Ignition Point 350 °C
Synonyms 2,2'-Methylenebis-(6-tert-butyl-4-methyl-phenol), Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)-methan
Hill Formula C23H32O2
Chemical Formula (2-(OH)-3-(CH3)3C-5-(CH3)C6H2)2CH2

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR