Methyl-4-formylbenzoat zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Methyl terephthalaldehydate, Methyl 4-formylbenzoate
M-Clarity™ Qualitätsstufe = MQ200
8.18490.0100
8.18490.0100EA
92.9
EUR
8.18490.0100
Methyl-4-formylbenzoat zur Synthese, Sigma-Aldrich®
Methyl-4-formylbenzoat
Formel:
C₉H₈O₃ Molecular Weight: 164.16 g/mol |
MDL:
MFCD00006950 CAS-Nummer: 1571-08-0 EINECS: 216-385-5 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 56 - 59 °C |
Molar Mass | 164.16 g/mol |
Bulk Density | 528 kg/m3 |
Boiling Point | 264 - 266 °C (1013 hPa) |
Flash Point | 128 °C |
Density | 1.2 g/cm3 (20 °C) |
Ignition Point | 240 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...