Methylpropionylacetat zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate

M-Clarity™ Qualitätsstufe = MQ200

8.18492.0010
8.18492.0010EA 62.1 EUR
8.18492.0010
Methylpropionylacetat zur Synthese, Sigma-Aldrich®
Methylpropionylacetat
Formel: C₆H₁₀O₃
Molecular Weight: 130,14 g/mol
MDL: MFCD00011705
CAS-Nummer: 30414-53-0
EINECS: 250-184-3

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) slightly soluble
Molar Mass 130.14 g/mol
Boiling Point 73 - 75 °C (20 hPa)
Density 1.04 g/cm3 (20 °C)
Synonyms 3-Oxopentanoic acid methyl ester
Hill Formula C6H10O3
Chemical Formula CH3CH2COCH2COOCH3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR