(4-Cyanbenzyl)triphenylphosphoniumchlorid zur Synthese
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.18511.0005
8.18511.0005EA
0
EUR
8.18511.0005
(4-Cyanbenzyl)triphenylphosphoniumchlorid zur Synthese
(4-Cyanbenzyl)triphenylphosphoniumchlorid
Formel:
C₂₆H₂₁ClNP |
MDL:
MFCD00011752 CAS-Nummer: 20430-33-5 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 326 - 338 °C |
Molar Mass | 413.88 g/mol |
Synonyms | Triphenyl-4-cyanobenzylphosphonium chloride |
Hill Formula | C26H21ClNP |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...