4'-Nitrozimtsäure zur Synthese
Lieferant: Merck
Synonyme:
trans-4-Nitrocinnamic acid, trans-3-(4-Nitrophenyl)propenoic acid
M-Clarity™ Qualitätsstufe = MQ200
8.18563.0025
8.18563.0025EA
106
EUR
8.18563.0025
4'-Nitrozimtsäure zur Synthese
4'-Nitrozimtsäure
Formel:
C₉H₇NO₄ Molecular Weight: 193,16 g/mol |
MDL:
MFCD00007381 CAS-Nummer: 882-06-4 EINECS: 212-924-3 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 288 - 291 °C |
Molar Mass | 193.15 g/mol |
Synonyms | trans-3-(4-Nitrophenyl)propenoic acid |
Hill Formula | C9H7NO4 |
Chemical Formula | 4-(NO2)C6H4CH=CHCOOH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...