(2R,3R)-(-)-2,3-O-Isopropyliden-D-threitol zur Synthese

Lieferant: Merck

Synonyme: (-)-2,3-O-Isopropylidene-D-threitol

8.18636.0001
8.18636.0001EA 0 EUR
8.18636.0001
(2R,3R)-(-)-2,3-O-Isopropyliden-D-threitol zur Synthese
(2R,3R)-(-)-2,3-O-Isopropyliden-D-threitol
Formel: C₇H₁₄O₄
Molecular Weight: 162,18 g/mol
MDL: MFCD00009761
CAS-Nummer: 73346-74-4
EINECS: 277-391-1

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 46 - 49 °C
Molar Mass 162.18 g/mol
Boiling Point 110 - 120 °C (1 hPa)
Flash Point 1.4574 (20 °C, 589 nm)
Density 1.15 g/cm3 (20 °C)
Synonyms (4R,5R)-(-)-4,5-Bis(hydroxymethyl)-2,2-dimethyl-1,3-dioxolane
Hill Formula C7H14O4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR