Kaliumperfluor-1-butansulfonat zur Synthese
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.18662.0050
8.18662.0050EA
157
EUR
8.18662.0050
Kaliumperfluor-1-butansulfonat zur Synthese
Kaliumperfluor-1-butansulfonat
Formel:
C₄F₉KO₃S |
MDL:
MFCD00007515 CAS-Nummer: 29420-49-3 EINECS: 249-616-3 |
Spezifikation Testergebnisse
Solubility | 51 g/l (25 °C) |
Melting Point | 264 - 266 °C |
Molar Mass | 338.19 g/mol |
Bulk Density | 1000 kg/m3 |
pH | 5.5 - 6.5 (50 g/l, H2O) |
Synonyms | Nonafluorobutane-1-sulfonic acid potassium salt |
Hill Formula | C4F9KO3S |
Chemical Formula | CF3(CF2)3SO3K |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...