Amylacetat ≥98,0% (durch GC), EMPLURA®, Supelco®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Formel:
C₇H₁₄O₂ Siedepunkt: 149 °C (1013 hPa) Schmelzpunkt: –71 °C Dichte: 0,876 g/cm³ (20 °C) Flammpunkt: 23 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009500 CAS-Nummer: 628-63-7 EINECS: 211-047-3 UN: 1104 ADR: 3,III |
|
Spezifikation Testergebnisse
Assay (GC, area%) | ≥ 98.0 % (a/a) |
Density (d 20 °C/ 4 °C) | 0.875 - 0.877 |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...