1-Hexadecanol zur Synthese, Sigma-Aldrich®
Lieferant: Merck
8.18704.0100
8.18704.1000
8.18704.0100EA
27.8
EUR
8.18704.0100
8.18704.1000
1-Hexadecanol zur Synthese, Sigma-Aldrich®
Spezifikation Testergebnisse
Solubility |
|
Melting Point | 46 - 52 °C |
Molar Mass | 242.44 g/mol |
Bulk Density | 410 - 450 kg/m3 |
Boiling Point | 305 - 330 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 155 °C DIN 51758 |
Density | 0.82 g/cm3 (50 °C) |
pH | (H2O) not applicable |
Ignition Point | 235 °C DIN 51794 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...